Assay Details
| Gene Symbol: LOC4351627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764931.2 | XP_015620417.1 | 22 - 23 | 2644 | 97 | |
| GenBank mRNA | AK058272.1 | - | 10 - 11 | 1090 | 97 | |
More Information
Set Membership: |
|

LOC4351627
Genomic Map