Assay Details
| Gene Symbol: LOC4352652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764610.2 | XP_015620096.1 | 1 - 2 | 276 | 95 | |
| GenBank mRNA | AK063508.1 | - | 1 - 2 | 259 | 95 | |
| AK071898.1 | - | 1 - 2 | 261 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015764610.2 transcripts.
Set Membership: |
|

LOC4352652
Genomic Map