Assay Details
| Gene Symbol: LOC4352652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764610.2 | XP_015620096.1 | 2 - 3 | 721 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352652| Gene Symbol: LOC4352652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764610.2 | XP_015620096.1 | 2 - 3 | 721 | 71 | |
Set Membership: |
|