Assay Details
| Gene Symbol: LOC4352652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764610.2 | XP_015620096.1 | 3 - 4 | 794 | 88 | |
| GenBank mRNA | AK071898.1 | - | 3 - 4 | 778 | 88 | |
More Information
Set Membership: |
|

LOC4352652
Genomic Map