Assay Details
Gene Symbol: LOC4352738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762795.2 | XP_015618281.1 | 2 - 3 | 445 | 127 | |
GenBank mRNA | AK108664.1 | - | 2 - 3 | 413 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
