Assay Details
Gene Symbol: LOC4352738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762795.2 | XP_015618281.1 | 3 - 4 | 599 | 118 | |
GenBank mRNA | AK108664.1 | - | 3 - 4 | 567 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
