Assay Details
Gene Symbol: LOC4352738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762795.2 | XP_015618281.1 | 5 - 6 | 759 | 113 | |
GenBank mRNA | AK108664.1 | - | 5 - 6 | 727 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
