Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 13 - 14 | 2008 | 72 | |
GenBank mRNA | AF251074.1 | - | 2 - 3 | 262 | 72 | |
AK061906.1 | - | 10 - 11 | 1262 | 72 | ||
AK065165.1 | - | 11 - 12 | 2052 | 72 | ||
AK121511.1 | - | 13 - 14 | 1970 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
