Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 14 - 15 | 2245 | 72 | |
GenBank mRNA | AF251074.1 | - | 3 - 4 | 499 | 72 | |
AK061906.1 | - | 11 - 12 | 1499 | 72 | ||
AK065165.1 | - | 12 - 13 | 2289 | 72 | ||
AK121511.1 | - | 14 - 15 | 2207 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
