Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 1 - 2 | 484 | 80 | |
GenBank mRNA | AK065165.1 | - | 1 - 2 | 370 | 80 | |
AK121511.1 | - | 1 - 2 | 446 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
