Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 2 - 3 | 599 | 67 | |
GenBank mRNA | AK065165.1 | - | 2 - 3 | 485 | 67 | |
AK121511.1 | - | 2 - 3 | 561 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
