Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 5 - 6 | 993 | 74 | |
GenBank mRNA | AK061906.1 | - | 2 - 3 | 247 | 74 | |
AK121511.1 | - | 5 - 6 | 955 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
