Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 6 - 7 | 1051 | 80 | |
GenBank mRNA | AK061906.1 | - | 3 - 4 | 305 | 80 | |
AK121511.1 | - | 6 - 7 | 1013 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
