Assay Details
Gene Symbol: LOC4352741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765102.2 | XP_015620588.1 | 9 - 10 | 1355 | 79 | |
GenBank mRNA | AK061906.1 | - | 6 - 7 | 609 | 79 | |
AK121511.1 | - | 9 - 10 | 1317 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
