Assay Details
Gene Symbol: LOC4352925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763858.2 | XP_015619344.1 | 8 - 9 | 2327 | 98 | |
GenBank mRNA | AK060641.1 | - | 1 - 2 | 138 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
