Assay Details
Gene Symbol: LOC4350138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761818.2 | XP_015617304.1 | 1 - 2 | 469 | 87 | |
GenBank mRNA | AK111558.1 | - | 1 - 2 | 273 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761818.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
