Assay Details
Gene Symbol: LOC4350138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761818.2 | XP_015617304.1 | 2 - 3 | 3104 | 113 | |
XM_015761819.2 | XP_015617305.1 | 3 - 4 | 3211 | 113 | ||
GenBank mRNA | AK105669.1 | - | 1 - 2 | 1962 | 113 | |
AK111558.1 | - | 2 - 3 | 2908 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
