Assay Details
| Gene Symbol: LOC4350275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762497.2 | XP_015617983.1 | 5 - 6 | 691 | 87 | |
| GenBank mRNA | AK103343.1 | - | 5 - 6 | 628 | 87 | |
More Information
Set Membership: |
|

LOC4350275
Genomic Map