Assay Details
| Gene Symbol: LOC4350686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759799.1 | XP_015615285.1 | 3 - 4 | 1098 | 148 | |
| XM_015759800.1 | XP_015615286.1 | 3 - 4 | 1111 | 148 | ||
| GenBank mRNA | AK064895.1 | - | 3 - 4 | 1060 | 148 | |
More Information
Set Membership: |
|

LOC4350686
Genomic Map