Assay Details
Gene Symbol: LOC4350702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760922.2 | XP_015616408.1 | 5 - 6 | 989 | 78 | |
GenBank mRNA | AK072007.1 | - | 5 - 6 | 567 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
