Assay Details
Gene Symbol: LOC4350702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760922.2 | XP_015616408.1 | 8 - 9 | 1263 | 130 | |
GenBank mRNA | AK072007.1 | - | 8 - 9 | 841 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
