Assay Details
Gene Symbol: LOC4351155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761786.2 | XP_015617272.1 | 6 - 7 | 2638 | 145 | |
XM_015761787.2 | XP_015617273.1 | 6 - 7 | 2635 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
