Assay Details
Gene Symbol: LOC4324413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765408.2 | XP_015620894.1 | 1 - 2 | 357 | 106 | |
GenBank mRNA | AK068747.1 | - | 1 - 2 | 334 | 106 | |
AK103737.1 | - | 1 - 2 | 316 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
