Assay Details
Gene Symbol: LOC4324413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765408.2 | XP_015620894.1 | 8 - 9 | 1344 | 75 | |
GenBank mRNA | AK068747.1 | - | 8 - 9 | 1321 | 75 | |
AK103737.1 | - | 8 - 9 | 1303 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
