Assay Details
Gene Symbol: LOC4327632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769105.2 | XP_015624591.1 | 3 - 4 | 387 | 102 | |
XM_015769115.2 | XP_015624601.1 | 3 - 4 | 387 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
