Assay Details
Gene Symbol: LOC4327632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769105.2 | XP_015624591.1 | 6 - 7 | 693 | 65 | |
XM_015769115.2 | XP_015624601.1 | 6 - 7 | 693 | 65 | ||
XM_015769127.2 | XP_015624613.1 | 5 - 6 | 561 | 65 | ||
XM_015769143.2 | XP_015624629.1 | 5 - 6 | 558 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
