Assay Details
| Gene Symbol: LOC4326767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762239.2 | XP_015617725.1 | 1 - 2 | 334 | 69 | |
| XM_015762248.2 | XP_015617734.1 | 1 - 2 | 332 | 69 | ||
| XM_015762257.2 | XP_015617743.1 | 1 - 2 | 331 | 69 | ||
| GenBank mRNA | AK243477.1 | - | 1 - 2 | 243 | 69 | |
More Information
Set Membership: |
|

LOC4326767
Genomic Map