Assay Details
Gene Symbol: LOC4324284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766009.2 | XP_015621495.1 | 1 - 2 | 128 | 106 | |
GenBank mRNA | AK065256.1 | - | 1 - 2 | 103 | 106 | |
BT129299.1 | - | 101 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766009.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
