Assay Details
Gene Symbol: LOC4324284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766009.2 | XP_015621495.1 | 3 - 4 | 366 | 83 | |
GenBank mRNA | AK065256.1 | - | 3 - 4 | 341 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
