Assay Details
Gene Symbol: LOC4327785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766948.2 | XP_015622434.1 | 6 - 7 | 742 | 79 | |
GenBank mRNA | AK064007.1 | - | 6 - 7 | 562 | 79 | |
AK102521.1 | - | 6 - 7 | 596 | 79 | ||
EU847008.1 | - | 6 - 7 | 577 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
