Assay Details
Gene Symbol: LOC4326991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766135.2 | XP_015621621.1 | 5 - 6 | 754 | 103 | |
GenBank mRNA | AK121409.1 | - | 5 - 6 | 734 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
