Assay Details
| Gene Symbol: LOC4326806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767029.2 | XP_015622515.1 | 9 - 10 | 1052 | 82 | |
| GenBank mRNA | AK066111.1 | - | 9 - 10 | 1059 | 82 | |
More Information
Set Membership: |
|

LOC4326806
Genomic Map