Assay Details
Gene Symbol: LOC4327659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755112.1 | XP_015610598.1 | 2 - 3 | 400 | 71 | |
XM_015755759.1 | XP_015611245.1 | 2 - 3 | 347 | 71 | ||
GenBank mRNA | AK069698.1 | - | 1 - 2 | 737 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
