Assay Details
| Gene Symbol: LOC4326617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766409.2 | XP_015621895.1 | 16 - 17 | 2305 | 59 | |
| GenBank mRNA | AK060394.1 | - | 6 - 7 | 753 | 59 | |
| AK068431.1 | - | 7 - 8 | 806 | 59 | ||
More Information
Set Membership: |
|

LOC4326617
Genomic Map