Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 14 - 15 | 1439 | 70 | |
| XM_015757449.2 | XP_015612935.1 | 14 - 15 | 1302 | 70 | ||
| GenBank mRNA | AB110983.1 | - | 14 - 15 | 1093 | 70 | |
| AK102997.1 | - | 14 - 15 | 1278 | 70 | ||
More Information
Set Membership: |
|

LOC4324139
Genomic Map