Assay Details
Gene Symbol: LOC4324139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757442.2 | XP_015612928.1 | 4 - 5 | 662 | 103 | |
XM_015757449.2 | XP_015612935.1 | 4 - 5 | 525 | 103 | ||
GenBank mRNA | AB110983.1 | - | 4 - 5 | 316 | 103 | |
AK102997.1 | - | 4 - 5 | 501 | 103 |
Gene Symbol: LOC9272129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780549.2 | XP_015636035.1 | 2 - 3 | 841 | 103 | |
GenBank mRNA | AK064202.1 | - | 2 - 3 | 638 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780549.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
