Assay Details
Gene Symbol: LOC4324139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757442.2 | XP_015612928.1 | 5 - 6 | 719 | 81 | |
XM_015757449.2 | XP_015612935.1 | 5 - 6 | 582 | 81 | ||
GenBank mRNA | AB110983.1 | - | 5 - 6 | 373 | 81 | |
AK102997.1 | - | 5 - 6 | 558 | 81 |
Gene Symbol: LOC9272129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780549.2 | XP_015636035.1 | 3 - 4 | 898 | 81 | |
GenBank mRNA | AK064202.1 | - | 3 - 4 | 695 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780549.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
