Assay Details
| Gene Symbol: LOC4324328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769999.2 | XP_015625485.1 | 4 - 5 | 330 | 89 | |
| XM_015770109.2 | XP_015625595.1 | 3 - 4 | 266 | 89 | ||
| GenBank mRNA | AK103859.1 | - | 4 - 5 | 309 | 89 | |
| BT129192.1 | - | 4 - 5 | 315 | 89 | ||
| BT131367.1 | - | 3 - 4 | 261 | 89 | ||
More Information
Set Membership: |
|

LOC4324328
Genomic Map