Assay Details
Gene Symbol: LOC4324328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769999.2 | XP_015625485.1 | 5 - 6 | 429 | 106 | |
XM_015770109.2 | XP_015625595.1 | 4 - 5 | 365 | 106 | ||
GenBank mRNA | AK103859.1 | - | 5 - 6 | 408 | 106 | |
BT129192.1 | - | 5 - 6 | 414 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
