Assay Details
| Gene Symbol: LOC4326960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766701.2 | XP_015622187.1 | 13 - 14 | 1475 | 80 | |
| GenBank mRNA | AK120714.1 | - | 13 - 14 | 1482 | 80 | |
| BT129140.1 | - | 2 - 3 | 150 | 80 | ||
More Information
Set Membership: |
|

LOC4326960
Genomic Map