Assay Details
Gene Symbol: LOC4327977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771949.2 | XP_015627435.1 | 3 - 4 | 1006 | 65 | |
XM_015771950.2 | XP_015627436.1 | 4 - 5 | 976 | 65 | ||
XM_015771951.2 | XP_015627437.1 | 3 - 4 | 1006 | 65 | ||
XM_015771952.2 | XP_015627438.1 | 4 - 5 | 976 | 65 | ||
GenBank mRNA | AK074023.1 | - | 4 - 5 | 967 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
