Assay Details
Gene Symbol: LOC4327977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771949.2 | XP_015627435.1 | 5 - 6 | 1206 | 68 | |
XM_015771950.2 | XP_015627436.1 | 6 - 7 | 1176 | 68 | ||
XM_015771951.2 | XP_015627437.1 | 5 - 6 | 1206 | 68 | ||
XM_015771952.2 | XP_015627438.1 | 6 - 7 | 1176 | 68 | ||
XM_015771953.2 | XP_015627439.1 | 4 - 5 | 1184 | 68 | ||
GenBank mRNA | AK074023.1 | - | 6 - 7 | 1167 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
