Assay Details
| Gene Symbol: LOC4328152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770673.2 | XP_015626159.1 | 11 - 12 | 1093 | 80 | |
| GenBank mRNA | AK121932.1 | - | 11 - 12 | 1074 | 80 | |
More Information
Set Membership: |
|

LOC4328152
Genomic Map