Assay Details
| Gene Symbol: LOC4328152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770672.2 | XP_015626158.1 | 4 - 5 | 597 | 114 | |
| XM_015770673.2 | XP_015626159.1 | 4 - 5 | 597 | 114 | ||
| XM_015770674.2 | XP_015626160.1 | 4 - 5 | 597 | 114 | ||
| XM_015770675.2 | XP_015626161.1 | 4 - 5 | 597 | 114 | ||
| GenBank mRNA | AK121932.1 | - | 4 - 5 | 578 | 114 | |
More Information
Set Membership: |
|

LOC4328152
Genomic Map