Assay Details
Gene Symbol: LOC4328221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767828.2 | XP_015623314.1 | 1 - 2 | 729 | 73 | |
GenBank mRNA | AK108836.1 | - | 2 - 3 | 625 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
