Assay Details
Gene Symbol: LOC4328481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768595.2 | XP_015624081.1 | 4 - 5 | 535 | 61 | |
XM_015768597.2 | XP_015624083.1 | 4 - 5 | 535 | 61 | ||
XM_015768598.2 | XP_015624084.1 | 4 - 5 | 535 | 61 | ||
XM_015768599.2 | XP_015624085.1 | 4 - 5 | 535 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
