Assay Details
Gene Symbol: LOC4328481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768595.2 | XP_015624081.1 | 5 - 6 | 680 | 142 | |
XM_015768597.2 | XP_015624083.1 | 5 - 6 | 680 | 142 | ||
XM_015768598.2 | XP_015624084.1 | 5 - 6 | 680 | 142 | ||
XM_015768599.2 | XP_015624085.1 | 5 - 6 | 680 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
