Assay Details
| Gene Symbol: LOC4328481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768595.2 | XP_015624081.1 | 6 - 7 | 781 | 85 | |
| XM_015768597.2 | XP_015624083.1 | 6 - 7 | 781 | 85 | ||
| XM_015768598.2 | XP_015624084.1 | 6 - 7 | 781 | 85 | ||
| XM_015768599.2 | XP_015624085.1 | 6 - 7 | 781 | 85 | ||
More Information
Set Membership: |
|

LOC4328481
Genomic Map