Assay Details
Gene Symbol: LOC4329036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768497.2 | XP_015623983.1 | 2 - 3 | 365 | 70 | |
GenBank mRNA | AK058613.1 | - | 2 - 3 | 319 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
