Assay Details
| Gene Symbol: LOC4329468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767064.2 | XP_015622550.1 | 5 - 6 | 818 | 69 | |
| XM_015767065.2 | XP_015622551.1 | 5 - 6 | 815 | 69 | ||
| GenBank mRNA | AK064485.1 | - | 3 - 4 | 304 | 69 | |
| DQ286558.1 | - | 4 - 5 | 459 | 69 | ||
More Information
Set Membership: |
|

LOC4329468
Genomic Map